Merck
Z742013
Duran® Youtility bottle, GL 45, complete
amber glass, capacity 500 mL
Synonym(e):
Duran® Youtility
Größe auswählen
4 EA
€ 217,00
€ 217,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
4 EA
€ 217,00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
€ 217,00
Check Cart for Availability
Materialien
amber glass
borosilicate glass 3.3 cap
round bottle
Verbindung
Joints threaded neck (GL45)
Sterilität
non-sterile
Leistungsmerkmale
graduations
with cap
Verpackung
pack of 4 ea
Hersteller/Markenname
DURAN® 218864457
Kapazität
500 mL
Allgemeine Beschreibung
  • Specially shaped gripping zones on both sides of the bottle for easier and safer handling
  • Opening or closing of the bottle is faster, and fully compatible with DIN GL 45 closures and other accessories
  • Slimmer YOUTILITY bottle shape allows a more optimal use of limited space in autoclaves and laboratory refrigerators
  • Pre-defined labelling area is compatible with DURAN self-adhesive YOUTILITY bottle labels
  • Nominal volume is shown at the top of the easy-to-read graduation scale for fast determination of the volumes
  • Sequentially sized bottles are available in four volumes
  • Moulded from the DURAN borosilicate 3.3 pharmacopeia type 1 neutral glass
  • Retrace Code for full product traceability
  • Each bottle is supplied as a complete system, with a pouring ring (cyan polypropylene) and a GL 45 cap (cyan polypropylene)
Leistungsmerkmale und Vorteile
  • UV protection: USP <660> (Spectral Transmission) and EP (3.2.1) compliant. Less than 10% spectral transmission between 290 nm and 450 nm.
  • Manufactured from ambered DURAN® borosilicate glass 3.3. DURAN® is a neutral glass, classified as a Glass Type I according to USP <660>, EP (3.2.1) and JP <7.01>, and meets ASTM E438, Type I, Class A requirements
  • The only external application of the colour ensures that internal surfaces have standard DURAN® glass properties. Very durable and chemically resistant amber colour.
  • Easier and safer handling with the award winning design. Specially shaped gripping zones on both sides of the bottle.
  • Significantly faster opening or closing with the shortened bottle thread which remains fully compatible with GL 45 closures and other accessories
  • The optimized, ergonomic shape of the screw cap has comfortable grooves and ridges for a more efficient and easier tightening or removal, especially with smaller sized or gloved hands.
  • The pre-defined labelling areas are compatible with the dedicated DURAN® self-adhesive YOUTILITY bottle labels for unambigious bottle identification.
  • The slimmer bottle shape allows a more optimal use of limited space in autoclaves and laboratory refrigerators.
  • The tried and tested cap plug sealing with sealing lip system ensures a liquid tight seal.
  • Award winning product design: Winner of seven international awards: Red Dot 2014, iF 2014, Silver IDEA International Excellence Award 2014, GOOD DESIGN AWARD Japan 2014, Special Mention German Design award 2015, Nominert DESIGNPREIS DER BUNDESREPUBLIC DEUTSCHLAND 2015, Silver DDC "Good Design 15" Award
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)