Merck
Z693774
LabJaws hook lattice rod connector
stainless steel
Synonym(e):
clamp holders, frame connectors
About This Item
UNSPSC-Code:
41100000
NACRES:
NB.35
Materialien
stainless steel
Hersteller/Markenname
Troemner 916358
Backenkapazität
0-13 mm
Allgemeine Beschreibung
Corrosion resistant. The hook allows for one-handed assembly of two components with one adjustment screw.
Rechtliche Hinweise
LabJaws is a trademark of Henry Troemner LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)