Merck
Z693456
2mag MIX 15 eco multi-position magnetic stirrer
AC/DC input 230 V AC, Schuko plug
Synonym(e):
laboratory stirrers, magnetic stir plates, magnetic stirrers, stirrers
Größe auswählen
1 EA
€ 1.200,00
€ 1.200,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 1.200,00
About This Item
UNSPSC-Code:
41103806
NACRES:
NB.72
€ 1.200,00
Check Cart for Availability
Materialien
stainless steel plate
Leistungsmerkmale
CE compliant
Verpackung
pack of 1 ea
Hersteller/Markenname
2mag 30115
Parameter
120 to 1200 rpm speed (stirring)
Platte B × L
245 nm × 425 mm
Rührervolumen
1-1500 mL , per stirring position
Allgemeine Beschreibung
Ultra flat, wear-free magnetic stirrer with 15 stirring positions, up to 1,500 mL stirring volume suitable for 15 x 250 mL beaker glasses.
Price-optimized magnetic stirrer with 15 stirring positions, 100% maintenance-free and wear-free by inductive 2mag-Magnetic-Drive concept for powerful mixing of solutions, speed range from 120 to 1,200 rpm, 100% synchronized speeds, jerk-free stirring even at low speeds.
Clear digital display for stirrer speed settings, SoftStart procedure for reliable catching/centering and safe acceleration of the stirring bar. Automatic memory function to save the last settings, outstanding long-life cycle of the hermetically sealed drive system, resistant stainless steel housing for easy and fast cleaning.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)