Merck
Z682713
Ace drying flask, pear-shaped, with side indents,
capacity 500 mL, joint: ST/NS 24/40, plastic-coated glass
Synonym(e):
Aldrich® drying flask, pear-shaped, with side indents
Größe auswählen
1 EA
€ 187,00
€ 187,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 187,00
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
€ 187,00
Check Cart for Availability
Materialien
pear-shaped flask
plastic-coated glass
single-neck
Verbindung
joint: ST/NS24/40
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
case of 1
Hersteller/Markenname
Ace Glass 3994-120
Kapazität
500 mL
Allgemeine Beschreibung
These pear shaped drying flasks for rotary evaporators have side indents which increase the mixing action when the flask is rotated on an evaporator .
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)