Merck
Z680419
Duran® GL 45 connection system caps and accessories
GL 45 blue PP screw cap with 2 hose connections, 6-9 mm
Synonym(e):
293102807, Connection System Accessories, Duran Connection Caps, GL 45 Connection System
Größe auswählen
2 EA
€ 66,80
€ 66,80
Check Cart for Availability
Größe auswählen
Ansicht ändern
2 EA
€ 66,80
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.17
€ 66,80
Check Cart for Availability
Materialien
blue-gray polypropylene
Hersteller/Markenname
DURAN® 293102807
Parameter
-40-140 °C temp. limit
Allgemeine Beschreibung
GL 45 screw cap with either two or three ports that have GL 14 thread. This is a flexible modular system which has four different hose diameters available: 1.6 mm, 3.0 mm, 3.2 mm and 6.0 mm that can be connected.
Sterile pressure equalization is possible through use of membrane filter.
Leistungsmerkmale und Vorteile
  • Manufactured from PP, EPDM
  • Highly versatile as the screw cap fits the standard GL 45 thread
  • 2-port hose connectors allow easy connection of silicone tubing to lab bottles for liquid transfer
  • EPDM ensures a gas tight seal with bottle
  • Grey polypropylene cap center rotates freely, allowing the bottle to be unscrewed without the need to disconnect the tubing
  • Autoclavable at 121 or 134 °C
  • Temperature resistant up to +140 °C
  • Lab-washer-proof
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)