Merck
Z678783
FORTUNA® Universal Optifix® HF dispenser
dispensing volume 1-5 mL
Größe auswählen
1 EA
€ 935,00
€ 935,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 935,00
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
€ 935,00
Check Cart for Availability
Hersteller/Markenname
Fortuna Graf 101.106-33
dispensing volume measuring range
1-5 mL
Volumen
1-5 mL
Fitting
GL 40
GL 45
Verwandte Kategorien
Allgemeine Beschreibung
For all liquids that can be handled with the Optifix dispensers and for hydrofluoric acid (HF)
Features:
  • Dispensing cylinder made of PTFE
  • Massive PTFE encased dispensing piston
  • FIX adapter made of PTFE
  • Ergonomic finger protection cap
  • Fast and easy volume setting with additional fine adjustment
  • Valve block made of PTFE, with additional safety valve in the ventilation tube, no metal springs in the valves - no corrosion
Supplied:
FORTUNA Universal Optifix HF dispenser, with 2 PTFE adapters GL40 + GL45 & 1 PTFE intake tube
Rechtliche Hinweise
FORTUNA is a registered trademark of Poulten & Graf Ltd.
Optifix is a registered trademark of Poulten & Graf Ltd.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)