Merck
Z568538
Sigma-Aldrich
Aldrich® solvent storage/dispensing flask, septum-inlet, with PTFE inlet valve
joint: ST/NS 24/40, capacity 100 mL
Synonym(e):
solvent dispenser bottles
Größe auswählen
1 EA
€ 443,00
€ 443,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 443,00
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
€ 443,00
Check Cart for Availability
Materialien
borosilicate glass
round bottom flask
single-neck
Verbindung
joint: ST/NS24/40
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
pack of 1
Hersteller/Markenname
Sigma-Aldrich
Kapazität
100 mL
Allgemeine Beschreibung
Designed for use with PureSolv Micro systems with a standard taper dispensing joint. PTFE inlet valve may be closed after filling to isolate dry solvent and to remove flask from system and take to another location for use. Use septum Z565695 or Z565679.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)