Merck
Z553867
Sigma-Aldrich
Microwave Synthesis: Chemistry at the Speed of Light
About This Item
ISBN-10:
0-9722229-0-1
ISBN-13:
978-0-9722229-0-7
UNSPSC-Code:
55101509
Informationen zur Veröffentlichung
B. Hayes, CEM Publishing, 2003, 289 pp., soft cover
Allgemeine Beschreibung
Benefiting both the practicing chemist and student alike, this book discusses microwave-based chemistry for the organic laboratory. Topics include optimizing reactions, applications in microwave synthesis, atmospheric and pressurized reactions, choosing the best solvent for a microwave-assisted reaction, solvent-free reactions, and the fundamentals of microwave theory.
Inhaltsverzeichnis
Chapter 1 Introduction to Microwave Chemistry
Chapter 2 Solvents
Chapter 3 Chemical Reactions in the Presence and Absence of Solvents
Chapter 4 Synthetic Applications
Chapter 5 Getting Started with Microwave Synthesis
Chapter 6 Microwave Safety Considerations
Chapter 7 Microwave Hardware
Afterword
Glossary
References
Indexx
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)