Merck
Z516856
Sigma-Aldrich
Aldrich® two-neck round-bottom flask
capacity 100 mL, Joints: ST/NS 24/40 (2)
Synonym(e):
bottom flask, flask round, round flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
round bottom flask
two-neck
Verbindung
Joints: ST/NS24/40 (2)
Sterilität
non-sterile
Leistungsmerkmale
closure type Standard Taper Joint
Verpackung
pack of 1
Hersteller/Markenname
Sigma-Aldrich
Kapazität
100 mL
Allgemeine Beschreibung
Angled ST joints. Z516929 and Z516953 have 24/40 side joints.
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC