Merck
Z508527
Ace flat-bottom reaction flasks
capacity 4,000 mL, w/ O-ring groove
Synonym(e):
bottom flask, flat flask, reaction flasks
Größe auswählen
1 EA
€ 442,00
€ 442,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 442,00
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
€ 442,00
Check Cart for Availability
Materialien
borosilicate glass
flat bottom
single-neck
Sterilität
non-sterile
Leistungsmerkmale
closure type Lid
Verpackung
pack of 1
Hersteller/Markenname
Ace Glass 6511-60
Kapazität
4,000 mL
Allgemeine Beschreibung
Cylindrical heavy-wall reaction flask with a flat bottom rounded into side of wall, and extra-thick flat flange. No constriction at top facilitates introduction or removal of material and allows easy cleaning. Optional O-ring groove allows use of CAPFE O-rings rather than gaskets. Flask is supplied with one CAPFE O-ring. Order reaction flask heads and clamps separately.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)