Merck
Z502642
Sigma-Aldrich
Threaded vial connector
8-425 thread for 8-425
Größe auswählen
1 EA
€ 26,70
€ 26,70
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 26,70
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
€ 26,70
Check Cart for Availability
Fitting
8-425 thread for 8-425
Allgemeine Beschreibung
These threaded connectors provide a convenient method to connect standard screw thread sample vials to glassware with GPI threads and to standard taper joint adapters.
• Excellent for collecting samples from rotary evaporator traps
• Made of glass filled nylon with inert PTFE inner seals
• Temp. range: up to 200 °C
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)