Merck
Z414093
Sigma-Aldrich
Aldrich® offset enlarging adapter
female joint: ST/NS 24/40, male joint: ST/NS 14/20
Synonym(e):
glass adapter, glass adapters
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
Verbindung
female joint: ST/NS24/40
male joint: ST/NS14/20
Rechtliche Hinweise
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)