Merck
Z279897
Sigma-Aldrich
Norprene® A-60-G Laborschlauch
I.D. × O.D. 1/16 in. × 3/16 in.
Synonym(e):
Flexible Tubing, Saint Gobain AFL00003, Tygon A-60-G
Größe auswählen
50 FT
€ 69,00
€ 69,00
Voraussichtliches Versanddatum30. April 2025
Größe auswählen
Ansicht ändern
50 FT
€ 69,00
About This Item
UNSPSC-Code:
41105108
NACRES:
NB.35
€ 69,00
Voraussichtliches Versanddatum30. April 2025
Verpackung
pkg of 50 feet
Hersteller/Markenname
Tygon
ID × AD
1/16 in. × 3/16 in.
Wandabmessungen
1/16 in.
Allgemeine Beschreibung
This chemical pump tubing is a thermoplastic elastomer tubing with chemical compatibilities approaching fluoroelastomers and silicone tubing. Black, rubber-like tubing is heat sealable. Max. temp.: 275°F (135°C).
  • Chemical resistance to oxidizing agents, acids, and alkalies
  • Resistance to flex-fatigue, will not weaken or crack
Rechtliche Hinweise
Norprene is a registered trademark of Saint-Gobain Abrasives, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)