Merck
Z260223
Sigma-Aldrich
FORTUNA® Optifix® Safety bottle-top dispenser
Safety model, volume 6-30 mL
Größe auswählen
1 EA
€ 716,00
€ 716,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 716,00
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
€ 716,00
Check Cart for Availability
Leistungsmerkmale
subdivision 0.5 mL
Hersteller/Markenname
Fortuna Graf 101.107-44
Messbereich Volumen
6-30 mL
Volumen
6-30 mL
Fitting
GL 40
GL 45
Verwandte Kategorien
Allgemeine Beschreibung
For dispensing strong acids and aggressive chemicals
Features:
  • PTFE encased dispensing piston with sealing lip at its end
  • FIX adapter made of PTFE
  • Valve block made of PTFE, no metal springs in the valve - no corrosion
  • Visible dispensing process by the use of a dispensing cylinder made of glass, coated with plastic-protection against breakage
  • Ergonmic finger protection cap
  • Fast and easy volume setting with additional fine adjustment
Supplied as Optifix® Safety dispenser, 2 PTFE adapters GL40 + GL45 & 1 PTFE intake tube
Rechtliche Hinweise
FORTUNA is a registered trademark of Poulten & Graf Ltd.
Optifix is a registered trademark of Poulten & Graf Ltd.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)