Merck
SYNS620008
Synthware 180 degree high vacuum valve with protected o-rings
size 0-8 mm
Synonym(e):
Synthware S620008
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware S620008
Verwandte Kategorien
Allgemeine Beschreibung
With two 9 mm O.D. sidearms sealed to the barrel at a 180°angle to one another. Viton® O-ring on PTFE shaft are protected with precision PTFE wipers. The plug has a machined-in integral PTFE tip O-ring to make the primary seal. This design provides for an excellent vacuum tight seal without exposing any material other than PTFE to chemicals.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)