Merck
SYNS580008
Synthware 3-way high vacuum valve
size 0-8 mm
Synonym(e):
Synthware S580008
Größe auswählen
1 EA
€ 187,00
€ 187,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 187,00
About This Item
UNSPSC-Code:
40161501
NACRES:
NB.51
€ 187,00
Check Cart for Availability
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware S580008
Allgemeine Beschreibung
With two 9 mm O.D. sidearms sealed to the barrel at a 90° angle to one another. The non-rotating PTFE plug has three O-rings, the tip O-ring making the primary seal on the precision formed tapered seat.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)