Merck
SYNF661410
Synthware Erlenmeyer flask
joint: ST/NS 14/20, capacity 10 mL
Synonym(e):
Synthware F661410, glass conical flask
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.51
Materialien
flat bottom
glass flask
Verbindung
joint neck14/20
joint: ST/NS14/20
Sterilität
non-sterile
Leistungsmerkmale
without cap
Verpackung
pkg of 1 ea
Kapazität
10 mL
Allgemeine Beschreibung
With joint. Stopper is not supplied. All sizes including 1L flasks are hand-blown from tubing to ensure heavy and uniform wall.
Sonstige Hinweise
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.