Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 22,10 |
Über diesen Artikel
€ 22,10
material
black Nylon cap, flat bottom, glass flask
joint
joint neck14/20, joint: ST/NS14/10
sterility
non-sterile
feature
with cap
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F501010
capacity
10 mL
General description
Other Notes
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description capacity 10 mL, joint: ST/NS 14/10 | description capacity 25 mL, joint: ST/NS 14/10 | description capacity 50 mL, joint: ST/NS 19/22 | description capacity 50 mL, joint: ST/NS 24/40 |
| manufacturer/tradename Synthware F501010 | manufacturer/tradename Synthware F501025 | manufacturer/tradename Synthware F661950 | manufacturer/tradename Synthware F662450 |
| sterility non-sterile | sterility non-sterile | sterility non-sterile | sterility non-sterile |
| capacity 10 mL | capacity 25 mL | capacity 50 mL | capacity 50 mL |
| material glass flask, black Nylon cap, flat bottom | material glass flask, flat bottom, black Nylon cap | material flat bottom, glass flask | material flat bottom, glass flask |
| feature with cap | feature with cap | feature without cap | feature without cap |
| joint joint neck14/20, joint: ST/NS14/10 | joint joint neck14/20, joint: ST/NS14/10 | joint joint neck19/22, joint: ST/NS19/22 | joint joint neck24/40, joint: ST/NS24/40 |
Still not finding the right product?
Explore all of our products under Synthware™ Erlenmeyer flask
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?