Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 76,50 |
Über diesen Artikel
€ 76,50
material
borosilicate glass flask, round bottom , three-neck (vertical side necks)
joint
center joint: ST/NS19/22, side joint: ST/NS19/22
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F449900
capacity
100 mL, 100 mL
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description 100 mL, center joint: ST/NS 19/22, side joint: ST/NS 19/22 | description 100 mL, center joint: ST/NS 24/40, side joint: ST/NS 19/22 | description 100 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20 | description 100 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40 |
| capacity 100 mL , 100 mL | capacity 100 mL, 100 mL | capacity 100 mL , 100 mL | capacity 100 mL, 100 mL |
| manufacturer/tradename Synthware F449900 | manufacturer/tradename Synthware F444900 | manufacturer/tradename Synthware F441100 | manufacturer/tradename Synthware F444400 |
| material borosilicate glass flask, three-neck (vertical side necks), round bottom | material borosilicate glass flask, round bottom , three-neck (vertical side necks) | material round bottom , borosilicate glass flask, three-neck (vertical side necks) | material three-neck (vertical side necks), borosilicate glass flask, round bottom |
| joint side joint: ST/NS19/22, center joint: ST/NS19/22 | joint side joint: ST/NS19/22, center joint: ST/NS24/40 | joint side joint: ST/NS14/20, center joint: ST/NS14/20 | joint side joint: ST/NS24/40, center joint: ST/NS24/40 |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ three neck round bottom flask with vertical side necks
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?