Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 67,60 |
Über diesen Artikel
€ 67,60
material
borosilicate glass flask, round bottom , three-neck (vertical side necks)
joint
center joint: ST/NS14/20, side joint: ST/NS14/20
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F441150
capacity
50 mL, 50 mL
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description 50 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20 | description 500 mL, center joint: ST/NS 14/20, side joint: ST/NS 14/20 | description 250 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40 | description 500 mL, center joint: ST/NS 29/42, side joint: ST/NS 24/40 |
| manufacturer/tradename Synthware F441150 | manufacturer/tradename Synthware F441500 | manufacturer/tradename Synthware F444250 | manufacturer/tradename Synthware F446500 |
| capacity 50 mL, 50 mL | capacity 500 mL , 500 mL | capacity 250 mL, 250 mL | capacity 500 mL , 500 mL |
| joint side joint: ST/NS14/20, center joint: ST/NS14/20 | joint side joint: ST/NS14/20, center joint: ST/NS14/20 | joint side joint: ST/NS24/40, center joint: ST/NS24/40 | joint center joint: ST/NS29/42, side joint: ST/NS24/40 |
| material round bottom , three-neck (vertical side necks), borosilicate glass flask | material borosilicate glass flask, round bottom , three-neck (vertical side necks) | material three-neck (vertical side necks), round bottom , borosilicate glass flask | material three-neck (vertical side necks), borosilicate glass flask, round bottom |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ three neck round bottom flask with vertical side necks
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?