Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|---|---|
1 ea | Warenkorb auf Verfügbarkeit prüfen | € 34,30 |
Über diesen Artikel
€ 34,30
material
borosilicate glass flask, flat bottom , single-neck
joint
joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F124100
capacity
100 mL
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description capacity 100 mL, joint: ST/NS 24/40 | description capacity 300 mL, joint: ST/NS 24/40 | description capacity 500 mL, joint: ST/NS 24/40 | description 100 mL, joint: ST/NS 24/40 |
| capacity 100 mL | capacity 300 mL | capacity 500 mL | capacity 100 mL, 100 mL |
| manufacturer/tradename Synthware F124100 | manufacturer/tradename Synthware F124300 | manufacturer/tradename Synthware F124500 | manufacturer/tradename Synthware F304100 |
| joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 | joint joint: ST/NS24/40 |
| material flat bottom , borosilicate glass flask, single-neck | material single-neck , flat bottom , borosilicate glass flask | material borosilicate glass flask, single-neck , flat bottom | material borosilicate glass flask, single-neck , round bottom |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
Still not finding the right product?
Explore all of our products under Synthware™ single-neck flat-bottom flask
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?