Merck
SYNC102442
Synthware Vigreux distillation column
joint: ST/NS 24/40, L 200 mm
Synonym(e):
Synthware C102442, distilling column, fractional column
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.51
Verbindung
joint: ST/NS24/40
Verpackung
pkg of 1 ea
Hersteller/Markenname
Synthware C102442
Länge
200 mm
Verwandte Kategorien
Allgemeine Beschreibung
Column length is measured from the highest to lowest indent in the column.
Rechtliche Hinweise
Synthware is a trademark of Kemtech America Inc.