Merck
MT51107678
Mettler Toledo DX series ISE half-cells electrode
Copper selective
Größe auswählen
1 EA
€ 1.320,00
€ 1.320,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 1.320,00
About This Item
UNSPSC-Code:
39121436
NACRES:
NB.79
€ 1.320,00
Check Cart for Availability
Materialien
membrane (Solid state)
shaft (POM)
Hersteller/Markenname
Mettler-Toledo 51107678
Messbereich
1.0-1.0-4
Temperaturmessbereich
0-80 °C
pH-Wert
2-8
Allgemeine Beschreibung
Modular and versatile the METTLER TOLEDO DX series ion-selective half-cells consist of two elements: a universal shaft and an ion-specific membrane module. This module may be exchanged allowing you to measure the ion of your choice. Just order the membrane kit specific for that ion, mount the new module onto the shaft of your ISE half-cell, and you have a new ISE!
electrolyte for reference electrode: 1 mol/L KNO3, ISA solution: 1 mol/L KNO3
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)