Merck
MT30040000
Mettler-Toledo uMix magnetic stirrer, for use with SevenCompact and SevenExcellence
Must be connected to meter
Größe auswählen
1 EA
€ 323,00
€ 323,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 323,00
About This Item
UNSPSC-Code:
41103800
NACRES:
NB.79
€ 323,00
Check Cart for Availability
Hersteller/Markenname
Mettler-Toledo 30040000
Allgemeine Beschreibung
The uMix magnetic stirrer from Mettler Toledo provides an extra degree of precision for your applications. Powered and controlled externally, the magnetic stirrer allows for enhanced reproducibility of results and higher efficiency in the measuring process.
The magnetic stirrer can be combined with either the SevenCompact pH/Ion S220, the SevenCompact Conductivity S230, or the multiparameter SevenExcellence benchtop meters.
Rechtliche Hinweise
SevenCompact is a trademark of Mettler-Toledo, Inc.
SevenExcellence is a trademark of Mettler-Toledo, Inc.
uMix is a trademark of Mettler-Toledo, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)