Merck
MT30014097
Mettler Toledo InLab® trace conductivity probe
About This Item
UNSPSC-Code:
39121436
NACRES:
NB.79
Materialien
titanium shaft
Beschreibung
Mini-LTW connector
Hersteller/Markenname
Mettler-Toledo 30014097
Messbereich Leitfähigkeit
0.0001-1000 μS/cm
L × B
170 mm × 12 mm
Kabelgröße
1.8 m
Allgemeine Beschreibung
Low conductivity measurement requires special attention. When measuring pure or ultra-pure water there are several sources of potential error such as carbon dioxide from air and temperature inaccuracy: The digital conductivity sensor Mettler Toledo InLab® Trace has a high precision temperature probe. The conductivity and temperature signals are digitalized and combined through a special algorithm for unmatched conductivity accuracy.
The optional flow-cell made of polypropylene provides the perfect protection from carbon dioxide. Because it does not contain a single piece of conducting material, any interference is excluded.
For pure water we also offer a special temperature compensation mode to increase the reliability of your readings.
Each sensor is supplied with a certificate specifying the actual cell constant which makes calibration superfluous.
  • High precision, digitalized temperature and conductivity readings
  • Certified cell constant which makes calibration superfluous
Rechtliche Hinweise
InLab is a registered trademark of Mettler-Toledo, Inc.
Ähnliches Produkt
Produkt-Nr.
Beschreibung
Preisangaben
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)