Merck
GF61290907
Copper-beryllium alloy, Cu98Be2
foil, 0.5m coil, 0.005mm thickness, as rolled
Synonym(e):
CU070225
About This Item
UNSPSC-Code:
31152304
NACRES:
NA.23
Form
foil (coil - as rolled)
Hersteller/Markenname
Goodfellow 612-909-07
Größe × Dicke
0.5 m × 0.005 mm
Allgemeine Beschreibung
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)