GF36510601
Hastelloy® C276 - heat resisting alloy, Ni57Mo17Cr16FeWMn
foil, 150 x 150mm, 0.075mm thickness, hard, light tight (lt)
Synonym(e):
NI140271
Beschreibung
light tight (lt)
Form
foil (hard
hard)
Hersteller/Markenname
Goodfellow 365-106-01
Größe × Dicke
150 mm × 150 mm × 0.075 mm
Verwandte Kategorien
Allgemeine Beschreibung
For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow
Rechtliche Hinweise
Hastelloy is a registered trademark of Haynes International, Inc.
Hastelloy is a registered trademark of Haynes International, Inc.