Merck
DWK247866909
DURAN® desiccator with MOBILEX lid and screw thread closure
thread (DN 300)
Synonym(e):
247866909
Größe auswählen
1 EACH
€ 556,00
€ 556,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EACH
€ 556,00
About This Item
UNSPSC-Code:
41104510
NACRES:
NB.17
€ 556,00
Check Cart for Availability
Materialien
borosilicate glass 3.3
Verpackung
case of 1 ea
Hersteller/Markenname
DURAN® 247866909
Fitting
GL 32 cap
thread (DN 300)
Allgemeine Beschreibung
Due to the high wall-thickness of the vessels and the exact machining of the vacuum-tight ground joints on the lid and base, storage under vacuum is possible – even over extremely long periods. Suitable for full vacuum (0 mbar). Spare parts such as lids, bases, stopcocks and caps can be interchanged as required (observe DN). Available with or without porcelain plate. Easy-to-read scale with graduation marks to estimate the content. Fired-on, highly durable white print.
Rechtliche Hinweise
DURAN is a registered trademark of DWK Life Sciences
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)