Merck
DWK14396-250
KIMBLE® KIMAX® Media bottle
without cap, round transparent borosilicate glass, bottle capacity (250 mL), non-sterile
Größe auswählen
10 EA
€ 142,00
€ 142,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
€ 142,00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
€ 142,00
Check Cart for Availability
Materialien
round transparent borosilicate glass
Sterilität
non-sterile
Leistungsmerkmale
without cap
Verpackung
case of 10 ea
Hersteller/Markenname
Kimble® 14396-250
Flaschenkapazität
(250 mL)
Fitting
GL 45 neck
GL 45
Allgemeine Beschreibung
GL 45 Media Bottles ideal for general laboratory use including mixing, storing or transporting culture media, chemicals or solvents.
  • Enhanced graduations and marking spot made with chemically resistant white enamel paint
  • 30 mm ID opening
  • Autoclavable
  • Supplied without caps or with linerless GL 45 screw thread caps
  • Replaceable clear (natural) drip-free polypropylene pour ring is included on each bottle
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Leistungsmerkmale und Vorteile
  • Enhanced graduations and marking spot made with chemically resistant white enamel paint
  • 30 mm ID opening
  • Autoclavable
  • Supplied without caps or with linerless GL 45 screw thread caps
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Rechtliche Hinweise
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)