Merck
DWK14395-2000
KIMBLE® KIMAX® Media bottle
with cap, round transparent borosilicate glass bottle, bottle capacity (2,000 mL), non-sterile
Größe auswählen
4 EA
€ 262,00
€ 262,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
4 EA
€ 262,00
About This Item
UNSPSC-Code:
41121800
NACRES:
NB.43
€ 262,00
Check Cart for Availability
Materialien
blue polypropylene cap
round transparent borosilicate glass bottle
Sterilität
non-sterile
Leistungsmerkmale
with cap
Verpackung
case of 4 ea
Hersteller/Markenname
Kimble® 14395-2000
Flaschenkapazität
(2,000 mL)
Fitting
GL 45 neck
GL 45
Allgemeine Beschreibung
GL 45 Media Bottles ideal for general laboratory use including mixing, storing or transporting culture media, chemicals or solvents.
  • Enhanced graduations and marking spot made with chemically resistant white enamel paint
  • 30 mm ID opening
  • Autoclavable
  • Supplied without caps or with linerless GL 45 screw thread caps
  • Replaceable clear (natural) drip-free polypropylene pour ring is included on each bottle
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Leistungsmerkmale und Vorteile
  • Enhanced graduations and marking spot made with chemically resistant white enamel paint
  • 30 mm ID opening
  • Autoclavable
  • Supplied without caps or with linerless GL 45 screw thread caps
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Rechtliche Hinweise
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)