Merck
CLS65640200
PYREXPLUS® volumetric flask, with protective coating, Pyrex® stopper
capacity 200 mL
Synonym(e):
flasks glass, volumetric flasks, volumetric glass, Corning® 65640
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
borosilicate glass
long
Stopfengröße
13
Sterilität
non-sterile
Leistungsmerkmale
closure type ST stopper
Verpackung
case of 12
pack of 6
Hersteller/Markenname
Corning 65640-200
tol. measuring range
0.10 mL
Kapazität
200 mL
Allgemeine Beschreibung
Flask features a protective PVC coating for longer product life and safety. Protective coating helps prevent glass from shattering and reduces spills. Autoclavable (121 °C) and resistant to thermal shock.
Vorsicht
Do not place over direct heat or flame. Do not heat above 121 °C moist heat or 110 °C dry heat.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
PYREXPLUS is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)