Merck
CLS5564010
Pyrex® class A low-actinic volumetric flask, Pyrex® ST stopper
capacity 10 mL
Synonym(e):
flasks glass, volumetric flasks, volumetric glass
About This Item
UNSPSC-Code:
41121804
NACRES:
NB.43
Materialien
borosilicate glass
long
Verbindung
joint stopper9
Sterilität
non-sterile
Leistungsmerkmale
closure type ST stopper
Verpackung
case of 12
Hersteller/Markenname
Corning 55640-10
Corning 55640
tol. measuring range
0.02 mL
Kapazität
10 mL
Allgemeine Beschreibung
Low actinic stained glass provides protection for materials sensitive to light. The protective color is an integral part of flask, which retains mechanical strength, chemical stability, and thermal resistance of Pyrex brand labware.
Rechtliche Hinweise
PYREX is a registered trademark of Corning, Inc.