Merck
CLS1700100
Corning® coliform water test sample container, sterile
with sodium thiosulfate tablet, 120 mL polypropylene bottle, graduated, sterile, 100/cs
Synonym(e):
sterile sample bottles
Größe auswählen
100 EA
€ 92,30
€ 92,30
Check Cart for Availability
Größe auswählen
Ansicht ändern
100 EA
€ 92,30
About This Item
UNSPSC-Code:
41161502
NACRES:
NB.13
€ 92,30
Check Cart for Availability
Verpackung
case of 100
pack of 100
Hersteller/Markenname
Corning 1700-100
Verwandte Kategorien
Allgemeine Beschreibung
  • Well defined EPA Fill Line for 100 mL
  • Reinforced custody string
  • Large thumb tabs to reduce contamination and ease for opening
  • 2 thumb taps on either side of the locking mechanism
  • 1 in. headspace between water level and underside of closed lid
Leistungsmerkmale und Vorteile
  • Corning® coliform water test sample container, sterile, is used in the drinking water testing for the presence of coliform bacteria and other microbiological contaminants.
  • Pure polypropylene with Capacity: 120 mL total capacity.
  • Provided with a lid to reduce the chance of contamination.
  • Locking arrow assures sterility has not been compromised.
  • Inclusive of a sodium thiosulfate tablet in container.
  • Leak proof.
Rechtliche Hinweise
Corning is a registered trademark of Corning, Inc.
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)