Merck
BR90641
BRAND® glass beaker with spout, tall form
volume 400 mL, with graduation
Synonym(e):
Beakers, Glass
Größe auswählen
10 EA
€ 120,00
€ 120,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
€ 120,00
About This Item
UNSPSC-Code:
41121803
NACRES:
NB.43
€ 120,00
Check Cart for Availability
Materialien
borosilicate glass 3.3
Leistungsmerkmale
with graduation
Hersteller/Markenname
BRAND 90641
Volumen
400 mL
Allgemeine Beschreibung
The BRAND® Glass Beaker with Spout, tall form, is constructed from high-quality Boro 3.3 glass, ensuring durability and reliability in laboratory environments. Compliant with DIN EN ISO 3819 standards, this beaker features clear graduation markings for accurate liquid measurement. The tall form design provides increased capacity, while the integrated spout facilitates easy pouring, minimizing spills and enhancing precision during liquid transfer.
Sonstige Hinweise
Browse all glass beakers
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)