BR759240
BRAND® cap for UV-cuvette micro
blue
Synonym(e):
Accessories, Caps, Cuvettes, UV-Cuvettes
Empfohlene Produkte
Materialien
PE
blue
polyethylene
Verpackung
pack of 100 ea
Hersteller/Markenname
BRAND 759240
Verwandte Kategorien
Allgemeine Beschreibung
Round covers allow a tight closure and make it possible to store samples down to -20 °C.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG