Merck
BR705928
BRAND® Transferpette® S Multichannel Pipette
capacity (10-100 μL), 12-channel, adjustable volume, M12-100, DE-M
Synonym(e):
Pipettes, Piston-operated pipettes, Positive displacement principle, Repair set, Transferpettor
Größe auswählen
1 EACH
€ 815,00
€ 815,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EACH
€ 815,00
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
€ 815,00
Check Cart for Availability
Leistungsmerkmale
12-channel adjustable volume
Verpackung
case of 1 ea
Hersteller/Markenname
BRAND 705928
Kapazität
(10-100 μL)
Verwandte Kategorien
Allgemeine Beschreibung
The Transferpette S air displacement pipette allows you to work efficiently and ergonomically with both small and large volumes. Low operating forces and a short stroke ensure relaxed pipetting, even during long pipetting series. Due to its perfect ergonomics, the Transferpette S provides a comfortable grip in any hand position, whether you are right- or left-handed, or have big or small hands. Changing your grip is not necessary, since the microliter pipette provides true one-hand operation: set the volume, pipette, eject the tip.The volume-change protection is also easy to operate with gloves on and protects your set volume against accidental adjustment. The Transferpette S also features Easy Calibration technology, which makes calibration a breeze. Calibration can be carried out without tools. A compatibility table shows which tips fit which device, so that you always order the correct pipette tips.
Overview- Volume range of single-channel pipette: from 0.1 μl to 10 ml- Volume range of multi-channel pipette: from 0.5 μl to 300 μl- Short stroke and low operating forces- Universal tip cone- Completely autoclaving at 121 μC- One-hand operation for right- and left-handed users- Color code on volume adjustment wheel, shaft and volume-change protection- Easy insertion and ejection of tips due to O-rings on multi-channel pipettes- Can be calibrated easily and without tools thanks to Easy Calibration technology- Slim shaft for sampling from narrow vessels (centrifuge tubes)- Operating temperature from +15 μC to +40 μC- Vapor pressure up to 500 mbar- Viscosity up to 260 mPa s
Efficient: One-hand operation
Flexible: ergonomic design for all hand sizes
Reliable: volume-change protection prevents accidental adjustments
Leistungsmerkmale und Vorteile
  • Efficient: one-hand operation
  • Flexible: ergonomic design for all hand sizes
  • Reliable: volume-change protection prevents accidental adjustments
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)