Merck
BR704552
BRAND® seripettor® cap
for 2+10 mL
Synonym(e):
Accessories, Bottle-top Dispenser, Dispenser, seripettor®
Größe auswählen
1 EA
€ 20,50
€ 20,50
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 20,50
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
€ 20,50
Check Cart for Availability
Materialien
polypropylene
Beschreibung
cap for valve block
Verpackung
pack of 1 ea
Hersteller/Markenname
BRAND 704552
Verwandte Kategorien
Allgemeine Beschreibung
The BRAND® Seripettor® cap is a polypropylene (PP) cap designed to securely close the valve block of the Seripettor® system. Engineered for durability and reliability, this cap is suitable for autoclaving, ensuring it can withstand high temperatures and sterilization processes. Its precise fit helps maintain the integrity of the system, making it an essential component.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)