Merck
BR4600360
BRAND® Dispensette® S Digital bottle-top dispenser
volume range 5-50 mL, without recirculation valve
Synonym(e):
bottle top dispensers
Größe auswählen
1 EA
€ 556,00
€ 556,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 556,00
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
€ 556,00
Check Cart for Availability
Leistungsmerkmale
incremental 0.2 mL
DE-M marking
without recirculation valve
Hersteller/Markenname
BRAND 4600360
Volumenbereich
5-50 mL
Fitting
(GL 32-33)
GL 38
S 40
Allgemeine Beschreibung
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Leistungsmerkmale und Vorteile
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Sonstige Hinweise
Digital-The mechanical volume setting is easy to read and ensures accurate and reproducible volume control. Easy Calibration Technique for calibration adjustment in seconds without tools. A calibration flag indicates changes from factory settings.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Protokolle
This article provides testing instructions for BRAND Dispensette® S bottle-top liquid dispenser according to the standard ISO DIS 8655.
Verwandter Inhalt
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.