Merck
BR4600211
BRAND® Dispensette® S Fixed-volume bottle-top dispenser
volume 1 mL, with recirculation valve
Größe auswählen
1 EA
€ 404,00
€ 404,00
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 404,00
About This Item
UNSPSC-Code:
41121500
NACRES:
NB.01
€ 404,00
Check Cart for Availability
Leistungsmerkmale
DE-M marking
with recirculation valve
Hersteller/Markenname
BRAND 4600211
Volumen
1 mL
Fitting
(GL 28/S28)
GL 32
GL 38
S 40
Allgemeine Beschreibung
Dispensette S is suitable for an amazingly broad range of applications, particularly for many acids, saline solutions and organic solvents.
Leistungsmerkmale und Vorteile
  • Extremely easy filling and dispensing
  • Simple priming by optimized flow channels
  • Rapid and safe volume adjustment with internal scalloped track
Sonstige Hinweise
Fixed Volume- For standard applications. Simple calibration adjustment with a supplied tool.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)
Die Dokumentenbibliothek aufrufen
Verwandter Inhalt
Chemical compatibility chart for all Dispensette® S and Dispensette® S Organic models.