Größe auswählen
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
material
AR-Glass
packaging
pack of 12 ea
manufacturer/tradename
BRAND 27006
measuring range
accuracy: 0.02 mL
capacity
2.0 mL
General description
Legal Information
1 of 1
Dieser Artikel | |||
|---|---|---|---|
| description 2 mL with 0.02 mL graduations, not suitable for cotton plugs | description 10 mL with 0.1 mL graduations, suitable for cotton plugs | description 1 mL with 0.01 mL graduations, not suitable for cotton plugs | description 2 mL with 0.01 mL graduations, not suitable for cotton plugs |
| capacity 2.0 mL | capacity 10 mL | capacity 1.0 mL | capacity 2.0 mL |
| manufacturer/tradename BRAND 27006 | manufacturer/tradename BRAND 27011 | manufacturer/tradename BRAND 27002 | manufacturer/tradename BRAND 27005 |
| material AR-Glass | material AR-Glass | material AR-Glass | material AR-Glass |
| packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 12 ea |
| measuring range accuracy: 0.02 mL | measuring range accuracy: 0.1 mL | measuring range accuracy: 0.01 mL | measuring range accuracy: 0.01 mL |
Still not finding the right product?
Explore all of our products under BRAND® SILBERBRAND ETERNA Graduated pipette, Class B, Type 2
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?