Merck
BR152605
BRAND® tubing adapter
polypropylene, for tubing i.d., 3- 5 to 6-10 mm
Synonym(e):
Tubing, Tubing adapters
Größe auswählen
10 EA
€ 15,50
€ 15,50
Check Cart for Availability
Größe auswählen
Ansicht ändern
10 EA
€ 15,50
About This Item
UNSPSC-Code:
41121815
NACRES:
NB.35
€ 15,50
Check Cart for Availability
Materialien
polypropylene
Leistungsmerkmale
for tubing i.d. 3- 5 to 6-10 mm
Verpackung
pack of 10 ea
Hersteller/Markenname
BRAND 152605
Länge
55 mm
Allgemeine Beschreibung
The BRAND® tubing adapter is designed to provide a secure connection. Its translucent construction allows for easy visibility of fluid flow, facilitating monitoring during use.
Rechtliche Hinweise
BRAND is a registered trademark of BRAND GMBH + CO KG
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)