Merck
BAH369441010
SP Bel-Art® Sterileware Long Handled Sampling Spoon
white polystyrene, nominal capacity 4.93 mL , 1tsp, L × W 17.8 cm (7 in.) × 1.1875 in. (30 mm), sterile, pack of 10 ea (double wrapped)
Synonym(e):
Lab Scoop, dispensing tool, spoon
About This Item
UNSPSC-Code:
41122800
NACRES:
NB.35
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Materialien
white polystyrene
Sterilität
sterile
Verpackung
pack of 10 ea (double wrapped)
Hersteller/Markenname
Bel-Art H36944-1010
L × B
17.8 cm (7 in.) × 1.1875 in. (30 mm)
Nennkapazität
4.93 mL , 1tsp
Allgemeine Beschreibung
The long handle on these spoons enables users to reach into jars, bottles and other confined areas to minimize the potential of personal contact with sampled material.
Leistungsmerkmale und Vorteile
  • Ten individually packaged spoons are sealed in an outer bag to create multiples layers of protection against contaminants and debris
  • Ideal for use in multi-zone clean rooms
  • FDA grade polystyrene safe for food, drug and cosmetic use
Rechtliche Hinweise
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Sterileware is a trademark of SP Scienceware, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)