Fortfahren mit
BAF651000000
SP Bel-Art® Proculture Cordless Homogenizer unit for Micro-Tube Homogenizer System
includes cordless motor, 2 AA batteries and pestle adapter
Synonym(e):
651000000, F65100-0000, Homogenizer, grinding tool, hand held Homogenizer, hand held grinding tool
Anmelden zur Ansicht der Organisations- und Vertragspreise.
Über diesen Artikel
NACRES:
NB.11
UNSPSC Code:
41101504
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F65100-0000
battery power
2 AA
General description
Hand-held, cordless homogenizer features a simple push in / pull out pestle exchange and is ideal for thorough and faster processing of one to many samples. Matched pestle tips and micro tubes can be used either with the homogenizer or with hand processing.
Features and Benefits
  • Homogenize samples directly in their storage container
  • Obtain thorough mixtures with the tight pestle-to-microtube tolerance
  • Protect your samples—tubes and pestles are made from virgin polypropylene
Legal Information
Bel-Art is a registered trademark of Bel-Art Products, a division of SP
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)