Merck
BAF133820000
Wet surface pen
pack of 1 ea
Größe auswählen
1 EA
€ 47,40
€ 47,40
Check Cart for Availability
Größe auswählen
Ansicht ändern
1 EA
€ 47,40
About This Item
UNSPSC-Code:
41103032
NACRES:
NB.35
€ 47,40
Check Cart for Availability
Verpackung
pack of 1 ea
Hersteller/Markenname
Bel-Art F13382-0000
Allgemeine Beschreibung
Many other uses include changing information on wet labels, whether from water bath liquid nitrogen or a freezer. Filters can be marked while they are being processed under water-based buffers and lanes can be identified on wet blots before they are cut into strips for independent incubation with different antisera or probes. Alignment dots can be added to blotting filters placed over bacterial colonies or phage plaques on agar plates.
  • Utilize to make notes on wet surfaces
  • Will record data underwater
  • Mark slots on wet Southern, Northern and Western blots
Hier finden Sie alle aktuellen Versionen:
Analysenzertifikate (COA)