Ansicht ändern
| Größe/SKU | Verfügbarkeit | Preis |
|---|
Über diesen Artikel
NACRES:
NA.21
UNSPSC Code:
41111761
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Technischer Dienst
Benötigen Sie Hilfe? Unser Team von erfahrenen Wissenschaftlern ist für Sie da.
Unterstützung erhaltendescription
Electrode coating: Au 3 µin
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
concentration
substrate (FR4)
electrode surface area
1 cm2
size
49 × 16 × 0.5 mm
weight
0.767 g
greener alternative category
General description
Our flat square electrode has a surface area of 1cm2, it is coated with gold, chemically stable, biocompatible, and durable. Its main applications include: Energy material characterization; Sensors; Energy storage devices; Actuators; Material synthesis.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.
Still not finding the right product?
Explore all of our products under Electrode
Hier finden Sie alle aktuellen Versionen:
Besitzen Sie dieses Produkt bereits?