Skip to Content
MilliporeSigma
Triethanolamine analytical standard

Triethanolamine

Triethanolamine analytical standard
Synonym(s):
2,2′,2′′-Nitrilotriethanol, Tris(2-hydroxyethyl)amine, Triethanolamine
Linear Formula:
(HOCH2CH2)3N
CAS Number:
Molecular Weight:
149.19
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Feature
Formula Weight
Melting Point (°C)
pH Value
Physical Form
pK Value
Product Line
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique