Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

50926

Supelco

Triethanolamine

analytical standard

Synonym(s):

2,2′,2′′-Nitrilotriethanol, Tris(2-hydroxyethyl)amine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HOCH2CH2)3N
CAS Number:
Molecular Weight:
149.19
Beilstein:
1699263
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

5.14 (vs air)

vapor pressure

0.01 mmHg ( 20 °C)

Assay

≥99.0% (GC)

autoignition temp.

600 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

8.5 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.1% ethanolamine (GC)
≤0.2% water
≤0.3% diethanolamine (GC)

refractive index

n20/D 1.485 (lit.)

useful pH range

7.3-8.3

pKa (25 °C)

7.8

bp

190-193 °C/5 mmHg (lit.)

mp

17.9-21 °C (lit.)

solubility

soluble 149 g/L at 20 °C (completely)

density

1.124 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OCCN(CCO)CCO

InChI

1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2

InChI key

GSEJCLTVZPLZKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

354.2 °F - closed cup

Flash Point(C)

179 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imad J Matouk et al.
Biochimica et biophysica acta, 1843(7), 1414-1426 (2014-04-08)
The oncofetal H19 gene transcribes a long non-coding RNA(lncRNA) that is essential for tumor growth. Here we found that numerous established inducers of epithelial to mesenchymal transition(EMT) also induced H19/miR-675 expression. Both TGF-β and hypoxia concomitantly induced H19 and miR-675
Huan Zhang et al.
Reproduction (Cambridge, England), 148(1), 43-54 (2014-04-02)
In mammals, the primordial follicle pool, providing all oocytes available to a female throughout her reproductive life, is established perinatally. Dysregulation of primordial follicle assembly results in female reproductive diseases, such as premature ovarian insufficiency and infertility. Female mice lacking
Heidi I Chen et al.
Development (Cambridge, England), 141(23), 4500-4512 (2014-11-08)
Identifying coronary artery progenitors and their developmental pathways could inspire novel regenerative treatments for heart disease. Multiple sources of coronary vessels have been proposed, including the sinus venosus (SV), endocardium and proepicardium, but their relative contributions to the coronary circulation
Samuel V Mussi et al.
Pharmaceutical research, 31(8), 1882-1892 (2014-02-14)
To develop a nanostructured lipid carrier (NLC) co-loaded with doxorubicin and docosahexaenoic acid (DHA) and to evaluate its potential to overcome drug resistance and to increase antitumoral effect in MCF-7/Adr cancer cell line. The NLC was prepared by a hot
Francesco Trepiccione et al.
Kidney international, 86(4), 757-767 (2014-05-03)
Almost half of patients receiving lithium salts have nephrogenic diabetes insipidus. Chronic lithium exposure induces AQP2 downregulation and changes in the cellular composition of the collecting duct. In order to understand these pathophysiological events, we determined the earliest lithium targets

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service