Skip to Content
MilliporeSigma

1.37148

Triethanolamine

EMPROVE® EXPERT, Ph. Eur., JPE, NF

Synonym(s):

2,2′,2′′-Nitrilotriethanol, Tris(2-hydroxyethyl)amine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(HOCH2CH2)3N
CAS Number:
Molecular Weight:
149.19
UNSPSC Code:
12352104
Beilstein/REAXYS Number:
1699263
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

JPE, NF, Ph. Eur.

vapor density

5.14 (vs air)

vapor pressure

0.01 mmHg ( 20 °C)

product line

EMPROVE® EXPERT

form

liquid

autoignition temp.

600 °F

expl. lim.

8.5 %

technique(s)

API processing | cocrystal formation: suitable

refractive index

n20/D 1.485 (lit.)

useful pH range

7.3-8.3

pKa (25 °C)

7.8

bp

190-193 °C/5 mmHg (lit.)

mp

17.9-21 °C (lit.)

density

1.124 g/mL at 25 °C (lit.)

application(s)

liquid formulation
pharmaceutical
semi-solid formulation

storage temp.

2-25°C

SMILES string

OCCN(CCO)CCO

InChI

1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2

InChI key

GSEJCLTVZPLZKY-UHFFFAOYSA-N

General description

Finding the right excipient that matches your needs as well as regulatory demands can be a complicated challenge. With our application know-how and regulatory expertise, we support you in every step of development, scale-up, and production.
As part of our Emprove® Program, our raw materials are offered with extensive documentation facilitating compliance of your pharma and biopharma product, full supply chain transparency and risk mitigation. Our SAFC® portfolio of high-quality products for biopharmaceutical and pharmaceutical formulation and production withstands strict quality control procedures and is produced according to applicable cGMP guidelines.

Application

Triethanolamine (Trolamine) is a tertiary amine that acts as a basic buffer component, typically used as a final formulation buffer for small molecules pharmaceuticals. It may also be used as a process buffer in cell lysis and biomolecule downstream chromatography.
As a multi-compendial product, Triethanolamine (Trolamine) Emprove® Expert meets the N-nitrosodiethanolamine limits required by Ph Eur. Also, low microbial and endotoxin limits facilitate the use of this product in high-risk applications.

Legal Information

Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany
SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Triethanolamine


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

354.2 °F - closed cup

flash_point_c

179 °C - closed cup



Stay compliant with GMP using the Emprove® Program

Accelerate qualification, maintain compliance, and save time with Emprove® Dossiers.

Download Dossiers

Note: you will be brought to EmproveSuite.com. Your username and password are separate from SigmaAldrich.com and may differ.

Register for free and access Emprove® Dossiers within minutes. Upgrade your subscription plan to unlock advanced and customized content in the Emprove® Suite

Not finding what you are looking for? Visit the Emprove® Program Help page.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
137148602504054839022531