Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

1.08379

Millipore

Triethanolamine

GR for analysis

Synonym(s):

Triethanolamine, Tris(2-hydroxyethyl)amine, 2,2′,2′′-Trihydroxytriethylamine, TEA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
N(CH2CH2OH)3
CAS Number:
Molecular Weight:
149.19
MDL number:
UNSPSC Code:
12352104
EC Index Number:
203-049-8
NACRES:
NB.21
bp:
360 °C/1013 hPa (decomposition)
vapor pressure:
<0.01 hPa ( 20 °C)

vapor pressure

<0.01 hPa ( 20 °C)

Quality Level

Assay

≥99.0% (GC)

form

viscous liquid

autoignition temp.

325 °C (DIN 51794)

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

3.6-7.2 % (v/v)

dilution

(for analytical testing)

pH

10.5 (20 °C, 15 g/L in H2O)

bp

360 °C/1013 hPa (decomposition)

mp

21 °C

transition temp

flash point 190 °C

density

1.12 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

N(CCO)(CCO)CCO

InChI

1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2

InChI key

GSEJCLTVZPLZKY-UHFFFAOYSA-N

Application

  • Light energy utilization and microbial catalysis for enhanced biohydrogen: Ternary coupling system of triethanolamine-mediated Fe@C-Rhodobacter sphaeroides: Triethanolamine is used to enhance light energy utilization and microbial catalysis in a novel ternary system, significantly boosting biohydrogen production efficiency (Jiang et al., 2024).

  • Purification and use of crude green glycerol from the transesterification of triglycerides in the formulation of an alcohol gel hand sanitizer: Triethanolamine is used to optimize the formulation of a novel alcohol gel hand sanitizer, enhancing its microbial effectiveness and user safety (Trindade Mazala et al., 2024).

Analysis Note

Purity (GC): ≥ 99
Identity (IR-spectrum): conforms
Appearance: colorless to slightly yellow viscose liquid
Chloride (Cl): ≤ 0.0001 %
Sulfate (SO₄): ≤ 0.002 %
Heavy metals (as Pb): ≤ 0.0001 %
Fe (Iron): ≤ 0.0001 %
Ethanolamine (GC): ≤ 0.1 %
Diethanolamine (GC): ≤ 0.5 %
Sulfated ash (600 °C): ≤ 0.005 %
Water (according to Karl Fischer): ≤ 0.2 %

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

354.2 °F - closed cup

Flash Point(C)

179 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service