Skip to Content
MilliporeSigma
All Photos(1)

Documents

1615138

USP

Sodium trifluoroacetate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Sodium trifluoroacetate, Trifluoroacetic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COONa
CAS Number:
Molecular Weight:
136.01
Beilstein:
3597949
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

sodium trifluoroacetate

manufacturer/tradename

USP

mp

205-207 °C (dec.) (lit.)

density

1.49 g/mL (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].[O-]C(=O)C(F)(F)F

InChI

1S/C2HF3O2.Na/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1

InChI key

UYCAUPASBSROMS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

TFA/Trifluoroacetate is a common residual process impurity in the preparation of peptides or a counter ion in active pharmaceutical ingredients (API). Sodium trifluoroacetate USP Reference Standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Sodium trifluoroacetate reference standard intended for use in specified quality tests and assays.

Also used to prepare trifluoroacetic acid (TFA) Stock solution to determine the amount of trifluoroacetic acid (TFA) in peptides according to the given below general chapter of United States Pharmacopeia (USP):
  • 〈503.1〉 Trifluoroacetic acid (TFA) in peptides

Other Notes

This product is part of the USP Biologics program.
Sales restrictions may apply.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service